Learn More
Dodecamethylcyclohexasiloxane, TRC
Specifications
Specifications
| Chemical Name or Material | Dodecamethylcyclohexasiloxane |
| CAS | 540-97-6 |
| Purity Grade Notes | GC |
| Molecular Formula | C12 H36 O6 Si6 |
| Quantity | 1 g |
| Synonym | Cyclohexasiloxane, 2,2,4,4,6,6,8,8,10,10,12,12-dodecamethyl-, Cyclohexasiloxane, dodecamethyl- (6CI,7CI,8CI,9CI), 2,2,4,4,6,6,8,8,10,10,12,12-Dodecamethylcyclohexasiloxane, Cyclohexadimethylsiloxane, DC 246, DC 246 Fluid, Dodecamethylcyclohexasiloxane, Dodecamethylhexacyclosiloxane, Dow Corning 246, Dow Corning 246 Fluid, Hexadecamethylcyclohexasiloxane, SF 1258, SH 246, SH 246 Fluid, Xiameter PMX 0246 |
| InChI Formula | InChI=1S/C12H36O6Si6/c1-19(2)13-20(3,4)15-22(7,8)17-24(11,12)18-23(9,10)16-21(5,6)14-19/h1-12H3 |
| SMILES | C[Si]1(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O1 |
| IUPAC Name | 2,2,4,4,6,6,8,8,10,10,12,12-dodecamethyl-1,3,5,7,9,11-hexaoxa-2,4,6,8,10,12-hexasilacyclododecane |
| Molecular Weight (g/mol) | 444.92 |
| Show More |