Learn More
Vilbert Lourmat™ Docprint CX5 Gel Documentation System
Description
- For electrophoresis gels, autoradiographs, membranes
- Complete solution without PC
- Image transfer via USB key
- Self-contained system for simple documentation of gels
- Control unit with 8 in. screen for viewing images and thermoformed ABS camera support cone
- 12-bit scientific CCD camera (4096 levels of gray)
- Modulating exposure time, indication of saturation of the spots on the display
- Large 8 TFT LCD screen
- Save images to USB flash drive
- Possibility to connect directly a thermal printer or USB inkjet
- Software provided for editing, publication analysis: molecular weight calculation and optical density, inlay of image comments
Specifications
Specifications
| Model | Docprint CX5 Gel |
| Product Type | Gel Documentation System |
| Quantity | 1Each |