Learn More
DOCK1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5101550
Additional Details : Weight : 0.01000kg
Description
Antibody detects endogenous levels of total DOCK1.
This gene encodes a loosely associated peripheral membrane protein related to the LanC family of bacterial membrane-associated proteins involved in the biosynthesis of antimicrobial peptides. This protein may play a role as a peptide-modifying enzyme component in eukaryotic cells. Previously considered a member of the G-protein-coupled receptor superfamily, this protein is now in the LanC family. Multiple alternatively spliced variants, encoding the same protein, have been identified.Specifications
DOCK1 | |
Polyclonal | |
Unconjugated | |
DOCK1 | |
180 kDa protein downstream of CRK; 9130006G06Rik; AI854900; ced5; D630004B07Rik; dedicator of cytokinesis 1; dedicator of cyto-kinesis 1; dedicator of cytokinesis protein 1; dock 180; DOCK1; DOCK180; DOwnstream of CrK; RGD1566072; RP11-82L9.1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
1793, 309081, 330662 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q14185, Q8BUR4 | |
DOCK1 | |
A synthesized peptide derived from human DOCK1(Accession Q14185), corresponding to amino acid residues R1661-E1711. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |