Learn More
Tocris Bioscience™ Docetaxel
GREENER_CHOICE
Product Code. p-4817070
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 18782674
Brand: Tocris Bioscience™ 4056/10
Microtubule stabilizer
Binds and stabilizes microtubules; leads to cell cycle arrest and apoptosis. Displays significant antitumor activity in ovarian and gastric tumors. Prevents microtubule depolymerization and disassembly in the absence of GTP. Exhibits cytotoxic activity.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 114977-28-5
Molecular Formula C43H53NO14
Specifications
Quantity 10 mg
Formula Weight 807.88
Purity >99%
Molecular Weight (g/mol) 807.89
Chemical Name or Material Docetaxel