missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ DOC2A Recombinant Protein
335.00€ - 508.00€
Specifications
Accession Number | AAH63436 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer |
Gene ID (Entrez) | 8448 |
Molecular Weight (g/mol) | 69.74 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16112472
|
Abnova™
H00008448-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
16122472
|
Abnova™
H00008448-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
AAH63436 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
69.74 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DOC2A | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Protein Array, ELISA, Western Blot | |
8448 | |
DOC2A (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MRGRRGDRMTINIQEHMAINVCPGPIRPIRQISDYFPRGPGPEGGGGSGGEAPAHLVPLALAPPAALLGATTPEDGAEVDSYDSDDATALGTLEFDLLYDRASCTLHCSILRAKGLKPMDFNGLADPYVKLHLLPGACKANKLKTKTQRNTLNPVWNEDLTYSGITDDDITHKVLRIAVCDEDKLSHNEFIGEIRVPLRRLKPSQKKHFNICLERQVPLASPSSMSAALRGISCYLKELEQAEQGQGLLEERGRILLSLSYSSRRRGLLVGILRCAHLAAMDVNGYSDPYVKTYLRPDVDKKSKHKTCVKKKTLNPEFNEEFFYEIELSTLATKTLEVTVWDYDIGKSNDFIGGVSLGPGARGEARKHWSDCLQQPDAALERWHTLTSELPPAAGALSSA | |
DOC2A | |
Human | |
Recombinant | |
Solution |