missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAM-1/CD226 Mouse anti-Human, Alexa Fluor 700, Clone: 102511, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB666N-100UG
This item is not returnable.
View return policy
Description
DNAM-1/CD226 Monoclonal specifically detects DNAM-1/CD226 in Human samples. It is validated for Flow Cytometry.Specifications
| DNAM-1/CD226 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q15762 | |
| CD226 | |
| Mouse myeloma cell line NS0-derived recombinant human DNAM-1/CD226 Glu19-Asn247 (predicted) Accession # Q15762 | |
| RUO | |
| 10666 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 102511 | |
| Alexa Fluor 700 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CD226, CD226 antigenplatelet and T cell activation antigen 1, CD226 molecule, DNAM1, DNAM1adhesion glycoprotein, DNAM-1DNAX accessory molecule-1, DNAX accessory molecule 1, PTA1, T lineage-specific activation antigen 1 antigen, TLiSA1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human DNAM-1/CD226 in direct ELISAs and Western blots. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction