Learn More
STEMCELL Technologies DMH1
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
Description
- DMH1 (dorsomorphin homolog 1) is a selective inhibitor of activin receptor-like kinase 2 (ALK2; IC50 = 13 - 108 nM), a type I bone morphogenetic protein (BMP) receptor (Hao et al.; Mohedas et al.) DMH1 exhibits no detectable inhibition of ALK4, ALK5, AMPK, KDR (VEGFR2), or PDGFRβ, although it inhibits ALK1 and ALK3 at nanomolar concentrations (Hao et al.; Mohedas et al.)
Specifications
Specifications
| CAS | 1206711-16-1 |
| Synonym | Dorsomorphin homolog 1 |
| Quantity | 1 mg |
| Molecular Formula | C24H20N4O |
| Purity | ≥98% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.