missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Cells successfully cultured in DMEM include primary fibroblasts, neurons, glial cells, HUVECs, and smooth muscle cells
- Offers a variety of DMEM modifications for a range of cell culture applications
Specifications
Specifications
| Classification | Serum-free |
| Form | Liquid |
| Product Type | DMEM Cell Culture Media |
| Serum Level | Serum-free |
| Sterility | Sterile |
| Sterilization Method | Sterile-filtered |
| With Additives | Phenol Red, Sodium Bicarbonate, Glucose |
| Without Additives | No Glutamine, No Sodium Pyruvate |
| Quantity | 500 mL |
| Culture Type | Mammalian Cell Culture |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?