Learn More
STEMCELL Technologies DMEM with 1000 mg/L D-Glucose
Product Code. 17128091
Encompass
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17128091 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17128091 Supplier STEMCELL Technologies Supplier No. 36253
Dulbecco's Modified Eagle's Medium (DMEM) with 1000 mg/L D-glucose
  • Dulbecco's Modified Eagle's Medium (DMEM) with 1000 mg/L D­-Glucose is recommended for a wide variety of cell culture applications
  • Selection of suitable nutrient medium is dependent on type of cell, conditions of culture, and degree of chemical definition required for the cell culture application
Specifications
For Use With (Application) Cell Culture
Quantity 500 mL