Learn More
DL-Valine-3-d1, CDN
Product Code. 30321747
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30321747
Brand: CDN CDND29760.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material DL-Valine-3-D
CAS 79168-24-4
Molecular Formula C5 D H10 N O2
Quantity 0.1 g
Synonym Valine-3-d (9 CI, ACI), DL-Valine-3-d (ZCI), DL-(β-2 H)Valine, 2-Amino-3-methylbutanoic-3-d acid, DL-Valine-3-D
InChI Formula InChI=1 S/C5H11NO2/c1-3(2)4(6)5(7)8/h3-4 H,6H2,1-2H3,(H,7,8)/i3D
SMILES [2 H]C(C)(C)C(N)C(=O)O
IUPAC Name 2-amino-3-deuterio-3-methylbutanoic acid
Molecular Weight (g/mol) 118.1525
Formula Weight 118.0853 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity