Learn More
DL-Ornithine Hydrochloride, TRC
Product Code. 30425892
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30425892 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30425892 Supplier Toronto Research Chemicals Supplier No. TRCO695555500MG
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material DL-Ornithine Hydrochloride
CAS 1069-31-4
Molecular Formula C5H12N2O2 •HCl
Quantity 500 mg
Synonym DL-Ornithine Monohydrochloride, Ornithine Monohydrochloride, NSC 25163
InChI Formula InChI=1S/C5H12N2O2.ClH/c6-3-1-2-4(7)5(8)9;/h4H,1-3,6-7H2,(H,8,9);1H
SMILES OC(C(CCCN)N)=O.Cl
IUPAC Name 2,5-diaminopentanoic acid;hydrochloride
Molecular Weight (g/mol) 132.163646
Formula Weight 168.0666