Learn More
dl-Menthyl Myristate, TRC
Specifications
Specifications
| Chemical Name or Material | dl-Menthyl Myristate |
| CAS | 29066-37-3 |
| Molecular Formula | C24 H46 O2 |
| Quantity | 50 mg |
| Synonym | rel-(1R,2S,5R)-5-Methyl-2-(1-methylethyl)cyclohexyl Ester Tetradecanoic Acid, (±)-p-Menth-3-yl Ester Myristic Acid |
| InChI Formula | InChI=1S/C24H46O2/c1-5-6-7-8-9-10-11-12-13-14-15-16-24(25)26-23-19-21(4)17-18-22(23)20(2)3/h20-23H,5-19H2,1-4H3/t21-,22+,23-/m1/s1 |
| SMILES | CCCCCCCCCCCCCC(=O)O[C@@H]1C[C@H](C)CC[C@H]1C(C)C |
| IUPAC Name | [(1R,2S,5R)-5-methyl-2-propan-2-ylcyclohexyl] tetradecanoate |
| Molecular Weight (g/mol) | 366.621 |
| Formula Weight | 366.35 |