missing translation for 'onlineSavingsMsg'
Learn More

DL-Lysine Acetylsalicylate, TRC

Product Code. 30134210
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134210 1 g 1g
30007683 100 mg 100mg
30008613 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134210 Supplier Toronto Research Chemicals Supplier No. TRCL4885001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62952-06-1
Molecular Formula C9 H8 O4 . C6 H14 N2 O2
Molecular Weight (g/mol) 326.34
Synonym DL-Lysine acetylsalicylate, Lysine, 2-(acetyloxy)benzoate (1:1), DL-Lysine, mono[2-(acetyloxy)benzoate], Lysine, mono[2-(acetyloxy)benzoate] (9CI), Benzoic acid, 2-(acetyloxy)-, compd. with DL-lysine (1:1), Benzoic acid, 2-(acetyloxy)-, compd. with lysine (1:1), 2,6-diaminohexanoic acid compound with 2-acetoxybenzoic acid (1:1), Acelysin, Acelysine, Acetylsalicylic acid DL-lysine salt, Acetylsalicylic acid dl-lysine salt, Acetylsalicylic acid lysine salt, Asalis, Aspegic, Aspidol, Aspirin DL-lysine, Aspirin lysine salt, Aspirin-dl-lysine, Aspirisine, Aspisol, Atselizin, Cogla, DL-Lysine aspirin salt, Delgesic, Egicalm, LAS, Laboprin, Lysal, Lysine acetylsalicylate, Lysine monosalicylate acetate, Lysini racemici acetylsalicylase, Quinvet, Solpirin, Venopirin, Vetalgina, Vetalgine, o-Acetylsalicylic acid D,L-lysine salt
IUPAC Name 2-acetyloxybenzoic acid;2,6-diaminohexanoic acid
SMILES CC(=O)Oc1ccccc1C(=O)O.NCCCCC(N)C(=O)O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C9H8O4.C6H14N2O2/c1-6(10)13-8-5-3-2-4-7(8)9(11)12;7-4-2-1-3-5(8)6(9)10/h2-5H,1H3,(H,11,12);5H,1-4,7-8H2,(H,9,10)
Formula Weight 326.1478
Chemical Name or Material DL-Lysine Acetylsalicylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.