missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Leucine-d10-N-FMOC, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Specifications
Specifications
| Chemical Name or Material | N-[(9 H-Fluoren-9-ylmethoxy)carbonyl]-DL-leucine-2,3,3,4,5,5,5,5',5',5'-D10 |
| Molecular Formula | C21 D10 H13 N O4 |
| Quantity | 0.1 g |
| Synonym | N-[(9 H-Fluoren-9-ylmethoxy)carbonyl]-DL-leucine-2,3,3,4,5,5,5,5',5',5'-d10, DL-Leucine-d10-N-FMOC, 2-[[(9 H-Fluoren-9-ylmethoxy)carbonyl]amino]-4-(methyl-d3)pentanoic-2,3,3,4,5,5,5-d7 acid, N-9-Fluorenylmethoxycarbonyl-DL-leucine-d10, N-FMOC-DL-leucine-2,3,3,4,5,5,5,5',5',5'-d10, N-FMOC-DL-leucine-2,3,3,4,5,5,5,5',5',5'-d10 |
| InChI Formula | InChI=1 S/C21H23NO4/c1-13(2)11-19(20(23)24)22-21(25)26-12-18-16-9-5-3-7-14(16)15-8-4-6-10-17(15)18/h3-10,13,18-19 H,11-12H2,1-2H3,(H,22,25)(H,23,24)/i1D3,2D3,11D2,13 D,19 D |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])C([2 H])(NC(=O)OCC1c2ccccc2c3ccccc13)C(=O)O |
| IUPAC Name | 2,3,3,4,5,5,5-heptadeuterio-2-(9 H-fluoren-9-ylmethoxycarbonylamino)-4-(trideuteriomethyl)pentanoic acid |
| Molecular Weight (g/mol) | 363.4732 |
| Formula Weight | 363.2255 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction