missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Histidine-alpha,beta,beta-d3, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | DL-Histidine-alpha,beta,beta-d3 |
| CAS | 344299-50-9 |
| Molecular Formula | C6 D3 H6 N3 O2 |
| Quantity | 0.01 g |
| Synonym | DL-Histidine-α,β,β-D3, Histidine-α,β,β-d3 (9 CI) |
| InChI Formula | InChI=1 S/C6H9N3O2/c7-5(6(10)11)1-4-2-8-3-9-4/h2-3,5 H,1,7H2,(H,8,9)(H,10,11)/i1D2,5 D |
| SMILES | [2 H]C([2 H])(c1c[nH]cn1)C([2 H])(N)C(=O)O |
| IUPAC Name | 2-amino-2,3,3-trideuterio-3-(1 H-imidazol-4-yl)propanoic acid |
| Molecular Weight (g/mol) | 158.18 |
| Formula Weight | 158.0883 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction