missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Carnitine-d9 HCl (trimethyl-d9), CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | D,L Carnitine-d9 Chloride |
| CAS | 1219386-75-0 |
| Molecular Formula | C7 D9 H7 N O3 . Cl |
| Quantity | 0.05 g |
| Synonym | 3-Carboxy-2-hydroxy-N,N,N-(trimethyl-d9)-1-propanaminium Chloride, (3-Carboxy-2-hydroxypropyl)(trimethyl-d9)ammonium Chloride, Aplegin-d9, Bicarnesine-d9, (+/-)-Carnitine Chloride; |
| InChI Formula | InChI=1 S/C7H15NO3.ClH/c1-8(2,3)5-6(9)4-7(10)11;/h6,9 H,4-5H2,1-3H3;1 H/i1D3,2D3,3D3; |
| SMILES | [Cl-].[2 H]C([2 H])([2 H])[N+](CC(O)CC(=O)O)(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | (3-carboxy-2-hydroxypropyl)-tris(trideuteriomethyl)azanium;chloride |
| Molecular Weight (g/mol) | 206.72 |
| Formula Weight | 206.1384 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction