Learn More
DL-Aspartic-2,3,3-d3 Acid, CDN
Product Code. 30323832
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30323832
Brand: CDN CDND19530.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material DL-Aspartic-2,3,3-d3 Acid
CAS 14341-75-4
Molecular Formula C4 D3 H4 N O4
Quantity 0.5 g
Synonym (±)-Aspartic Acid-d3, Aminosuccinic Acid-d3
InChI Formula InChI=1 S/C4H7NO4/c5-2(4(8)9)1-3(6)7/h2H,1,5H2,(H,6,7)(H,8,9)/i1D2,2 D
SMILES [2 H]C([2 H])(C(=O)O)C([2 H])(N)C(=O)O
IUPAC Name 2-amino-2,3,3-trideuteriobutanedioic acid
Molecular Weight (g/mol) 136.12
Formula Weight 136.0563 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity