Learn More
Thermo Scientific Chemicals DL-Arginine, 98%
Product Code. p-7023702
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 11458137
Brand: Thermo Scientific Chemicals A17520.06
Request Bulk or Custom Format
CAS: 7200-25-1 | C6H14N4O2 | 174.204 g/mol
DL-Arginine is used in the synthesis of creatine and polyamines. DL-Arg is used in physicochemical analysis of amino acid complexation dynamics and crystal structure formations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
DL-Arginine is used in the synthesis of creatine and polyamines. DL-Arg is used in physicochemical analysis of amino acid complexation dynamics and crystal structure formations.
Solubility
Insoluble in water.
Notes
Air Sensitive. Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7200-25-1
Molecular Formula C6H14N4O2
Molecular Weight (g/mol) 174.204
MDL Number MFCD00063117
InChI Key ODKSFYDXXFIFQN-UHFFFAOYSA-N
Synonym dl-arginine, arginin, h-dl-arg-oh, 2-amino-5-carbamimidamidopentanoic acid, 2-amino-5-guanidinopentanoic acid, unii-fl26ntk3ep, dl-arginine, free base, fl26ntk3ep, wln: muyzm3yzvq &gh, arginine, dl
PubChem CID 232
ChEBI CHEBI:29016
IUPAC Name 2-amino-5-(diaminomethylideneamino)pentanoic acid
SMILES C(CC(C(=O)O)N)CN=C(N)N
Specifications
Melting Point ∼230°C (decomposition)
Beilstein 1725411
Solubility Information Insoluble in water.
Quantity 5 g
Formula Weight 174.2
Sensitivity Air Sensitive
Percent Purity 98%
Chemical Name or Material DL-Arginine
compliance-icons
Product Identifier
  • DL-Arginine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : DL-Arginine