Learn More
DL-6-Hydroxy Norleucine, TRC
Product Code. 30404073
Change view
Click to view available options
Quantity:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30404073 500 mg
30400007 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30404073 Supplier Toronto Research Chemicals Supplier No. TRCH948855500MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 305-77-1
Molecular Formula C6 H13 N O3
Molecular Weight (g/mol) 147.17
IUPAC Name 2-amino-6-hydroxyhexanoic acid
SMILES NC(CCCCO)C(=O)O
Specifications
InChI Formula InChI=1S/C6H13NO3/c7-5(6(9)10)3-1-2-4-8/h5,8H,1-4,7H2,(H,9,10)
Quantity 500 mg
Formula Weight 147.0895
Shipping Condition Room Temperature
Chemical Name or Material DL-6-Hydroxy Norleucine