Learn More
DL-4-Hydroxyphenyl-2,6-d2-alanine-2-d1, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | DL-Tyrosine-alpha,2,6-D3 |
| CAS | 73036-42-7 |
| Molecular Formula | C9 D3 H8 N O3 |
| Quantity | 0.25 g |
| Synonym | Tyrosine-α,2,6-d3 (9 CI), DL-Tyrosine-α,2,6-d3, DL-4-Hydroxyphenyl-2,6-d2-alanine-2-d1, DL-Tyrosine-α,2,6-D3, 2-Amino-3-(4-hydroxyphenyl-2,6-d2)propanoic-2-d acid, DL-Tyrosine-d3, DL-4-Hydroxyphenylalanine-d3 |
| InChI Formula | InChI=1 S/C9H11NO3/c10-8(9(12)13)5-6-1-3-7(11)4-2-6/h1-4,8,11 H,5,10H2,(H,12,13)/i1D,2 D,8 D |
| SMILES | [2 H]c1cc(O)cc([2 H])c1CC([2 H])(N)C(=O)O |
| IUPAC Name | 2-amino-2-deuterio-3-(2,6-dideuterio-4-hydroxyphenyl)propanoic acid |
| Molecular Weight (g/mol) | 184.207 |
| Formula Weight | 184.0927 g/mol |
| Show More |