missing translation for 'onlineSavingsMsg'
Learn More

DL-2-Bromohexanoic acid, 97%, Thermo Scientific™

Product Code. 10581851 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
This item is not returnable. View return policy

Product Code. 10581851

Brand: Thermo Scientific Acros 222121000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

Specifications

CAS 616-05-7
Melting Point 4°C
Color Brown
Density 1.3700g/mL
Boiling Point 136°C to 138°C (18.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C6H11BrO2
Linear Formula CH3(CH2)3CH(Br)CO2H
Refractive Index 1.4710 to 1.475
MDL Number MFCD00004218
Quantity 100 mL
Beilstein 02, 325
Merck Index 15, 1422
Specific Gravity 1.37
Synonym 2-bromocaproic acid, dl-2-bromohexanoic acid, hexanoic acid, 2-bromo, alpha-bromohexanoic acid, alpha-bromo-n-caproic acid, .alpha.-bromo-n-caproic acid, 2-bromo-hexanoic acid, .alpha.-bromocaproic acid, .alpha.-bromohexanoic acid, 2-bromo-n-hexanoic acid
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and ether
InChI Key HZTPKMIMXLTOSK-UHFFFAOYSA-N
SMILES CCCCC(C(=O)O)Br
IUPAC Name 2-bromohexanoic acid
Molecular Weight (g/mol) 195.06
PubChem CID 12013
Formula Weight 195.06
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material DL-2-Bromohexanoic acid, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • DL-2-Bromohexanoic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.