missing translation for 'onlineSavingsMsg'
Learn More

Dithio-2,2'-bis(N-methylbenzamide)-d6, TRC

Product Code. 30377754
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30377754 100 mg 100mg
30386043 250 mg 250mg
30381981 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30377754 Supplier Toronto Research Chemicals Supplier No. TRCD493952100MG
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dithio-2,2'-bis(N-methylbenzamide)-d6
Molecular Formula C16 D6 H10 N2 O2 S2
Quantity 100 mg
Synonym Densil P-d6
InChI Formula InChI=1S/C16H16N2O2S2/c1-17-15(19)11-7-3-5-9-13(11)21-22-14-10-6-4-8-12(14)16(20)18-2/h3-10H,1-2H3,(H,17,19)(H,18,20)/i1D3,2D3
SMILES [2H]C([2H])([2H])NC(=O)c1ccccc1SSc2ccccc2C(=O)NC([2H])([2H])[2H]
IUPAC Name N-(trideuteriomethyl)-2-[[2-(trideuteriomethylcarbamoyl)phenyl]disulfanyl]benzamide
Molecular Weight (g/mol) 338.477
Formula Weight 338.103
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.