Learn More
Lenz Laborglasintrumente Distilling Links with Splash Head
Product Code. 10335931
Click to view available options
Length (Metric):
150 mm
200 mm
300 mm
Unit Size:
Each
Product Code. 10335931
Brand: Lenz Laborglasintrumente 05043002
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • With two vertical NS cones and splash head
  • Made of DURAN™ tubing
Specifications
Length (Metric) 200 mm
Material Borosilicate Glass 3.3
Standard Taper Joint NS 19/26
Product Content Correction
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Politique de confidentialité.
Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis