Learn More
Lenz Laborglasintrumente Distilling Links with Splash Head
Product Code. 10244341
Change view
Click to view available options
Length (Metric):
150 mm
200 mm
300 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
10244341 150 mm Each
10020231 300 mm Each
10335931 200 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10244341 Supplier Lenz Laborglasintrumente Supplier No. 05043001
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • With two vertical NS cones and splash head
  • Made of DURAN™ tubing
Specifications
Length (Metric) 150 mm
Material Borosilicate Glass 3.3
Standard Taper Joint NS 14/23