Learn More
Lenz Laborglasinstrumente™ Distillation column to Hempel with fixed vacuum jacket socket/cone 29/32 800mm Lenz
Product Code. 10356021
Change view
Click to view available options
Standard Taper Joint:
29/32
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint unitSize
10356021 29/32 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10356021 Supplier Lenz Laborglasinstrumente™ Supplier No. 05119804
  • ONLINE EXCLUSIVE PRICE
    387.00€
    245.08€ / Pack of 1
    Save 141.92€
    36% Off
View all promotions
Estimated Shipment: 25-08-2026