Learn More
BOLA Distillation Apparata
Click to view available options
Capacity (Metric):
500 mL
1000 mL
2000 mL
4000 mL
6000 mL
Unit Size:
Each
Description
- Distillation Apparata of PTFE - with SAFE LAB-System consisting of: Reaction vessel
- flat flange (B281-03) Gasket
- flat flange (B 282-02) Lid
- flat flange (B 283-02) Joining piece
- flat flange (B 284-02) Dropping funnel
- 125 ml (B 285-01) Supporting device for thermometer (B 286-03)
- Thermometer for flasks (B 287-03)
- Stirrer bearing (B288-02)
- Moon-Shaped stirrer shaft (C 376-12)
- Stillhead 2x NS 29/32 (B 289-03)
- Distillation thermometer (B 290-03)
- Liebig condenser 270 mm (B 291-02)
- Receiver adapter (B 292-02)
- Vacuum stopcock (B 293-02)
- Round-bottom flask (A 158-06)
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| Dimensions (L x W x H) | 450 x 600 mm |
| Autoclavable | Autoclavable |