missing translation for 'onlineSavingsMsg'
Learn More

Chemetrics™ Dissolved Oxygen CHEMets™ Kit

Product Code. 11712403
Change view
Click to view available options
Range:
0 to 20 ppb
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range unitSize
11712403 0 to 20 ppb Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11712403 Supplier Chemetrics™ Supplier No. K7511

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Analytes Dissolved Oxygen
For Use With (Application) Monitoring dissolved oxygen in high-pressure boiler systems
Includes Refill, comparator, adhesive mounting clamp, permanent mounting clamp, sampling tube
No. of Tests 30
Range 0 to 20 ppb
Test Method Rhodazine D
compliance-icons compliance-icons
Product Identifier
  • CHEMets oxygen 0-20 PPB
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash all exposed externalbody areas thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P363-Wash contaminated clothing before reuse.
  • P405-Store locked up.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
  • MIXTURE LIST-Contain: diethylene glycol, potassium hydroxide,potassium pphosphate, monobasic
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.