Learn More
Rogo Sampaic™ 6-Piece Dissecting Set
Product Code. 11778531
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11778531 1 Ea. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11778531 Supplier Rogo Sampaic™ Supplier No. 55501010
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
A 6-piece dissection kit for general laboratory usage.
  • Provided in a case with a snap button closure
  • 1 pair of sharp scissors, rounded, 140mm
  • 1 pair of fine iris scissors, 110mm
  • 2 strong rounded forceps, 140mm
  • 1 fluted probe
Specifications
Product Type Dissecting Set
For Use With (Application) Dissection Applications
Quantity 1 Ea.