Learn More
Buerkle™ LaboPlast™ SteriPlast™ DispoPipette
Click to view available options
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 20
Pack of 50
Description
- Intended for sampling of low viscosity media
- All-layer samples can be obtained when the DispoPipette is inserted into a liquid while opened
- Target point samples are obtained by inserting while closed and opening the DispoPipette at the sampling point
- Two opening sizes (2.5 and 5.5mm) for different viscosities
- Clean room manufactured
- Sterile varieties are sterilized by gamma rays
Specifications
Specifications
| Product Type | Pipette |
| Material | HDPE |
| Capacity (Metric) | 200 mL |
| Disposable | Single-use |
| Sterility | Sterile |
| For Use With (Application) | For Taking Samples of Liquids Which Are Low in Viscosity |
| Length (Metric) | 1000 mm |