Learn More
Buerkle™ LaboPlast™ SteriPlast™ DispoPipette
GREENER_CHOICE
Product Code. 10767075
Click to view available options
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 20
Pack of 50
Product Code. 10767075
Brand: Buerkle™ 53935531
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Open and insert into liquid to obtain an all-layer sample is obtained. Or keep it closed to and then open once inserted up to the sampling point to obtain a target- point sample. Buerkle™ LaboPlast™ SteriPlast™ DispoPipette is very easy to operate.
  • Intended for sampling of low viscosity media
  • All-layer samples can be obtained when the DispoPipette is inserted into a liquid while opened
  • Target point samples are obtained by inserting while closed and opening the DispoPipette at the sampling point
  • Two opening sizes (2.5 and 5.5mm) for different viscosities
  • Clean room manufactured
  • Sterile varieties are sterilized by gamma rays
TRUSTED_SUSTAINABILITY
Specifications
Product Type Pipette
Material HDPE
Capacity (Metric) 200 mL
Disposable Single-use
Sterility Non-sterile
For Use With (Application) For Taking Samples of Liquids Which Are Low in Viscosity
Length (Metric) 1000 mm