Learn More
Buerkle™ Disposable Dropping Pipet
Shop All Bürkle Products
Click to view available options
Volume (Metric):
1.7 mL
1.8 mL
2.4 mL
Unit Size:
Pack of 100
Description
- LDPE
- dropping pipette, not graduated
Specifications
Specifications
| Material | LDPE |
| For Use With (Application) | Suited for Training Purposes, In Industrial Labs, During Field Research and Applications |
| Quantity | 1 Ea. |
| Volume (Metric) | 1.7 mL |
| Length (Metric) | 145 mm |
| Type | Dropping Pipette |
| Disposable | Single-use |