Learn More
HUBER Displacement Insert
Product Code. 10007852
Change view
Click to view available options
Length (Metric):
85 mm
124 mm
162 mm
172 mm
194 mm
245 mm
260 mm
265 mm
275 mm
For Use With (Equipment):
CC-205B
CC-308B
CC-315B
CC-410, CC-410 WL
CC510, CC515, CC905, CC905W, CC906W
CC510W,CC515W,CC520/W,CC525W,CC820/W
Ministat 125/230 W
Ministat 230/230 W
Ministat 240/240 W
Unit Size:
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) For Use With (Equipment) Unit Size
10007852 194 mm Ministat 240/240 W Each
10794616 124 mm CC-308B Each
10412142 172 mm Ministat 125/230 W Each
10407863 162 mm Ministat 230/230 W Each
10561073 245 mm CC510, CC515, CC905, CC905W, CC906W Each
10334475 260 mm CC-315B Each
10093772 265 mm CC510W,CC515W,CC520/W,CC525W,CC820/W Each
10781664 275 mm CC-410, CC-410 WL Each
10232635 85 mm CC-205B Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 10007852 Supplier HUBER Supplier No. 6820
  • ONLINE EXCLUSIVE PRICE
    627.00€
    576.67€ / Pack of 1
    Save 50.33€
    8% Off
 Restricted Item
Estimated Shipment: 12-11-2026
For use with refrigeration bath circulators
Displacement insert
Specifications
For Use With (Equipment) Ministat 240/240 W
Type Displacement Insert
Length (Metric) 194 mm