Learn More
Kinematica Dispersing Aggregate Dia. 5mm PT-DA 05/2EC-F078
Brand:  Kinematica 11030297
Dispersing Aggregate Dia. 5mm PT-DA 05/2EC-F078
Product Code. 15462082
  • ONLINE EXCLUSIVE PRICE
    1774.50€
    1641.00€ / Pack of 1
    Save 133.50€
    7% Off
Only null left
Estimated Shipment: 28-05-2026