Learn More
Kinematica Dispersing Aggregate Dia. 45mm PT-DA 45/6EC-F250
Brand:  Kinematica 11030329
Dispersing Aggregate Dia. 45mm PT-DA 45/6EC-F250
Product Code. 17570435
  • ONLINE EXCLUSIVE PRICE
    4877.25€
    4527.00€ / Each
    Save 350.25€
    7% Off