Learn More
Kinematica Dispersing Aggregate Dia. 30mm PT-DA 30/2ZEC-F250
Brand:  Kinematica 11030310
Dispersing Aggregate Dia. 30mm PT-DA 30/2ZEC-F250
Product Code. 15633616
  • ONLINE EXCLUSIVE PRICE
    3344.25€
    3104.00€ / Each
    Save 240.25€
    7% Off
Only null left
Estimated Shipment: 19-02-2026