Learn More
Kinematica Dispersing Aggregate Dia. 20mm PT-DA 20/2WEC-F193
Brand:  Kinematica 11030304
Dispersing Aggregate Dia. 20mm PT-DA 20/2WEC-F193
Product Code. 15697015
  • ONLINE EXCLUSIVE PRICE
    2661.75€
    2469.00€ / Each
    Save 192.75€
    7% Off
Only null left
Estimated Shipment: 02-02-2026