Learn More
Kinematica Dispersing Aggregate Dia. 50mm PT-DA 50/4EC-F250
Brand:  Kinematica 11030335
Dispersing Aggregate Dia. 50mm PT-DA 50/4EC-F250
Product Code. 16130351
  • ONLINE EXCLUSIVE PRICE
    4142.25€
    3841.00€ / Each
    Save 301.25€
    7% Off
Only null left
Estimated Shipment: 04-02-2026