Learn More
Kinematica Dispersing Aggregate Dia. 36mm PT-DA 36/4EC-F250
Brand:  Kinematica 11030317
Dispersing Aggregate Dia. 36mm PT-DA 36/4EC-F250
Product Code. 15576929
  • ONLINE EXCLUSIVE PRICE
    3738.00€
    3470.00€ / Pack of 1
    Save 268.00€
    7% Off
Only null left
Estimated Shipment: 11-05-2026