Learn More
Kartell™ Dispenser Dosipet HF
Product Code. 10151813
Change view
Click to view available options
Volume (Metric):
2.5 to 25 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10151813 2.5 to 25 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10151813 Supplier Kartell™ Supplier No. 8308100
  • ONLINE EXCLUSIVE PRICE
    1490.00€
    1412.08€ / Pack of 1
    Save 77.92€
    5% Off
Estimated Shipment: 09-12-2026
Dosing devices are designed for specific use with hydrofluoric acid solutions
  • The internal components that come into contact with these solutions are made with high quality and high resistance materials (PTFE and platinum-iridium), completely inert with hydrofluoric acid
  • Complete with PTFE piston
  • CE marked
  • IVD
  • Specifications
    Volume (Metric) 2.5 to 25 mL
    Increments 0.5 mL
    Model Dosipet HF-25
    Precision ± 0.15
    Thread Size 45 GL