missing translation for 'onlineSavingsMsg'
Learn More

Disopyramide, TRC

Product Code. 30383102
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30383102 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30383102 Supplier Toronto Research Chemicals Supplier No. TRCD493450100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Disopyramide
CAS 3737-09-5
Molecular Formula C21 H29 N3 O
Quantity 100 mg
Synonym Disopyramide, 2-Pyridineacetamide, α-[2-(diisopropylamino)ethyl]-α-phenyl- (7CI,8CI), α-[2-[Bis(1-methylethyl)amino]ethyl]-α-phenyl-2-pyridineacetamide, (RS)-disopyramide, (±)-Disopyramide, 4-Diisopropylamino-2-phenyl-2-(2-pyridyl)butyramide, Dicorantil, H 3292, Isorythm, Lispine, Ritmilen, Ritmodan, SC 7031, Searle 703, dl-Disopyramide, α-[2-(Diisopropylamino)ethyl]-α-phenyl-2-pyridineacetamide
InChI Formula InChI=1S/C21H29N3O/c1-16(2)24(17(3)4)15-13-21(20(22)25,18-10-6-5-7-11-18)19-12-8-9-14-23-19/h5-12,14,16-17H,13,15H2,1-4H3,(H2,22,25)
SMILES CC(C)N(CCC(C(=O)N)(c1ccccc1)c2ccccn2)C(C)C
IUPAC Name 4-[di(propan-2-yl)amino]-2-phenyl-2-pyridin-2-ylbutanamide
Molecular Weight (g/mol) 339.47
Formula Weight 339.2311
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.