Learn More
Disodium (RS)-2-Hydroxy-1,5-pentanedioate-2,3,3-d3,OD, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 2-Hydroxypentanedioic Acid-2 O,2,3,3-D4 Disodium Salt |
| CAS | 2483831-91-8 |
| Molecular Formula | C5 D4 H2 O5 . 2 Na |
| Quantity | 0.1 g |
| Synonym | Pentanedioic-2,3,3-d3 acid, 2-(hydroxy-d)-, sodium salt (1:2) (ACI), Disodium (RS)-2-Hydroxy-1,5-pentanedioate-2,3,3-d3,OD, 2-(Hydroxy-d)pentanedioic-2,3,3-d3 acid disodium salt, 2-Hydroxypentanedioic acid-2 O,2,3,3-D4 disodium salt, DL-α-Hydroxyglutaric acid-d4 disodium salt |
| InChI Formula | InChI=1 S/C5H8O5.2 Na/c6-3(5(9)10)1-2-4(7)8;;/h3,6 H,1-2H2,(H,7,8)(H,9,10);;/q;2*+1/p-2/i1D2,3 D,6 D;; |
| SMILES | [Na+].[Na+].[2 H]OC([2 H])(C(=O)[O-])C([2 H])([2 H])CC(=O)[O-] |
| IUPAC Name | disodium;2,3,3-trideuterio-2-deuteriooxypentanedioate |
| Molecular Weight (g/mol) | 196.1023 |
| Formula Weight | 196.0262 g/mol |
| Show More |