Learn More
Disodium Hydrogenphosphate EU Pharmacopoeia, Fisher Chemical™
Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.
Brand:  Fisher Chemical RP 072 1000
Additional Details : Weight : 1.00000kg
Product Code. 12900414
  • 112.00€ / 1000mL
Estimated Shipment: 13-06-2024
Specifications
Glass bottle
EU Ph Ref: 1033301
Disodium hydrogen phosphate solution.
1000 mL
Eu. Ph.
6988-21-2