Learn More
Virkon™ Disinfectant Virucidal Tablets
Product Code. 12318667
Change view
Click to view available options
Quantity:
10 Tablets/Tub, 20 Tubs/Cs.
50 Tablets/Tub, 12 Tubs/Cs.
Unit Size:
200 tablets
600 tablets
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12318667 50 Tablets/Tub, 12 Tubs/Cs. 600 tablets
12328667 10 Tablets/Tub, 20 Tubs/Cs. 200 tablets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12318667 Supplier Virkon™ Supplier No. 330013
  • ONLINE EXCLUSIVE PRICE
    497.00€
    428.37€ / 600 tablets
    Save 68.63€
    13% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Clean and disinfect work surfaces and equipment in a single, one-step operation. Virkon™ Disinfectant Virucidal Tablets provide the widest proven spectrum of any disinfectant.
Use biocides safely. Always read the label and product information before use.
  • Widest proven spectrum of any disinfectant - proven effective in independent tests against all 18 virus families affecting man and animals, including AIDS (HIV) and Hepatitis B
  • Excellent bactericide and fungicide - cleans and disinfects work surfaces and equipment in a single, one-step operation
  • Can be applied in powder form direct to spillages of blood and other body fluids
  • Easily dissolves to make a liquid disinfectant
  • Odourless, non-irritant to skin and eyes, non-toxic vapour phase, exceptionally low oral toxicity
  • Under COSHH requirements, no maximum exposure limit is required
  • Non-corrosive, non-bleaching and biodegradable
  • Any viral disease transmitted via surfaces or instruments contaminated with infected blood, body fluids, faeces or saliva can be controlled by correct application For cleaning and disinfecting work surfaces and equipment.
Specifications
Product Type Virucidal Disinfectant
Form Tablet
For Use With (Application) For Cleaning and Disinfecting Work Surfaces and Equipment
Quantity 50 Tablets/Tub, 12 Tubs/Cs.
Volume (Metric) 5 g/Tablet
compliance-icons
Product Identifier
  • relyon virkon tablets
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
  • P264-Wash skin thoroughly after handling.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: pentapotassium bis(peroxymonosulphate) bis(sulphate), sulphamidic acid, benzenesulphonic acid, potas