Learn More
KGW Isotherm™ Dish-Shaped Dewer Flasks, Aluminum Cover
Product Code. 10323241
Change view
Click to view available options
Capacity (Metric):
80 mL
120 mL
Diameter (Metric) Outer:
80-90 mm
95-110 mm
Height (Metric):
75 mm
90 mm
Model:
SCH 6 CAL
SCH 9 CAL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Diameter (Metric) Outer Height (Metric) Model unitSize
10323241 80 mL 80-90 mm 75 mm SCH 6 CAL Each
10179600 120 mL 95-110 mm 90 mm SCH 9 CAL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10323241 Supplier KGW Isotherm™ Supplier No. 1191
Only null left
Add to basket
Add to basket
Reliable and economical Dewar flasks according to DIN EN ISO 16496. KGW-Isotherm Dish-Shaped Dewer Flasks, Aluminum Cover pairs a bowl shaped glass flask in a protective casing made of stucco aluminum for easy handling. Lid sold separately.
  • Made of borosilicate glass according to ISO 3585
  • Structured aluminum cover: antimagnetic and stainless
  • Fully silvered
  • Applications:
    • tempering of round bottom flasks
    • cooling of small samples
    • suitable as a hot or cold bath
    • suitable on magnetic stirrer
  • Chemical characteristics:
    • Hydrolytic resistance: according to ISO 719 (98°C)
    • Hydrolytic resistance: according to ISO 720 (121°C)
    • Acid resistance: according to ISO 1776
    • Alkaline resistance: according to ISO 695-A2
  • Temperature range:
    • Glass refill from -200°C to + 150°C, for a short time 200°C
    • Casing with mounting rubber and Latizell buffer from -10°C to + 60°C
Specifications
Color Silver
Diameter (Metric) Inner Mouth 67 mm
Diameter (Metric) Outer 80-90 mm
For Use With (Application) Tempering, Cooling, Warming, Stirring
Height (Metric) Inner 40 mm
Height (Metric) 75 mm
Model SCH 6 CAL
Base Material Aluminum
Capacity (Metric) 80 mL
Material Borosilicate Glass
Style Fully silvered