Learn More
Dirithromycin, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 62013-04-1 |
| Molecular Formula | C42H78N2O14 |
| MDL Number | MFCD00865041 |
| Quantity | 1 g |
| SMILES | CC1[C@@]([C@](O)([C@H](OC([C@@H]2C)=O)CC)C)([H])O[C@H](COCCOC)N[C@@]1([H])[C@](C[C@@](O)([C@@H]([C@@H](C)[C@]2([H])O[C@@](O[C@@H](C)[C@@H]3O)([H])C[C@@]3(C)OC)O[C@@](O[C@H](C)C[C@@H]4N(C)C)([H])[C@@H]4O)C)([H])C |
| Molecular Weight (g/mol) | 835.07 |
| Percent Purity | 98% |
| Chemical Name or Material | Dirithromycin |