Learn More
Brady™ Directional Arrow Tape
Product Code. 12717946
Click to view available options
Dimensions (LxW):
33 m x 100 mm
33 m x 25 mm
33 m x 50 mm
Color:
Black on Gray
Black on Ochre
Black on Orange
Black on White
White on Black
White on Blue
White on Green
White on Purple
White on Red
Unit Size:
1 roll
Product Code. 12717946
Brand: Brady™ 275117
Directional flow arrow tape can enhance safety, improve operating effciency and provides 360 degree visibility.
  • Directional flow arrow tape can enhance safety, improve operating effciency and provides 360 degree visibility.
Specifications
Certifications/Compliance ROHS Environmental Compliance, EU Directive 2002/95/EC, European Directives 67/548 and 88/379
Description Directional Arrow Tape
Adhesive Type Permanent Acrylic, Permanent, Pressure-sensitive
Color Black on White
Finish Gloss
Format Tape
Material Polyester
Product Type Pipe Marker
For Use With (Application) Pipe and valve marking
No. per Pack 1300 Markers/Roll
Quantity 1300 Pk.
Temperature Range (English) -40°F to +230°F
Temperature Range (Metric) -40°C to +110°C
Thickness (English) 0.0039 in.
Thickness (Metric) 0.099 mm
Dimensions (LxW) 33 m x 50 mm
Length (English) 108.27 ft.
Length (Metric) 33 m
Width (English) 1.97 in.
Width (Metric) 50 mm
Alternate Part Number ARROWTAPE:WH/BLACKARROW-50MMX33M