missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dipyridamole Mono-O-Beta-D-glucuronide, TRC
Specifications
Specifications
| Chemical Name or Material | Dipyridamole Mono-O-beta-D-glucuronide |
| CAS | 63912-02-7 |
| Molecular Formula | C30 H48 N8 O10 |
| Quantity | 5 mg |
| Synonym | β-D-Glucopyranosiduronic acid, 2-[[6-[bis(2-hydroxyethyl)amino]-4,8-di-1-piperidinylpyrimido[5,4-d]pyrimidin-2-yl](2-hydroxyethyl)amino]ethyl, Glucopyranuronic acid, ester with 2,2',2'',2'''-[(4,8-dipiperidinopyrimido[5,4-d]pyrimidine-2,6-diyl)dinitrilo]tetraethanol (7CI), 2-[[6-[Bis(2-hydroxyethyl)amino]-4,8-di-1-piperidinylpyrimido[5,4-d]pyrimidin-2-yl](2-hydroxyethyl)amino]ethyl β-D-glucopyranosiduronic acid, Pyrimido[5,4-d]pyrimidine, β-D-glucopyranosiduronic acid deriv., Dipyridamole mono-O-β-D-glucuronide |
| InChI Formula | InChI=1S/C30H48N8O10/c39-15-11-37(12-16-40)29-31-19-20(25(33-29)35-7-3-1-4-8-35)32-30(34-26(19)36-9-5-2-6-10-36)38(13-17-41)14-18-47-28-23(44)21(42)22(43)24(48-28)27(45)46/h21-24,28,39-44H,1-18H2,(H,45,46)/t21-,22-,23+,24-,28+/m0/s1 |
| SMILES | OCCN(CCO)c1nc(N2CCCCC2)c3nc(nc(N4CCCCC4)c3n1)N(CCO)CCO[C@@H]5O[C@@H]([C@@H](O)[C@H](O)[C@H]5O)C(=O)O |
| IUPAC Name | (2S,3S,4S,5R,6R)-6-[2-[[6-[bis(2-hydroxyethyl)amino]-4,8-di(piperidin-1-yl)pyrimido[5,4-d]pyrimidin-2-yl]-(2-hydroxyethyl)amino]ethoxy]-3,4,5-trihydroxyoxane-2-carboxylic acid |
| Molecular Weight (g/mol) | 680.75 |
| Formula Weight | 680.3493 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?