Learn More
Esculape Laboratoire™ Diphoterine Eye Wash
Product Code. 13418786
Change view
Click to view available options
Volume (Metric):
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
13418786 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13418786 Supplier Esculape Laboratoire™ Supplier No. 8205020
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Use Esculape Laboratoire™ Diphoterine Eye Wash to limit the extent of the burns and lesions by restricting the extent and severity of the lesions.
  • Sterile
  • Ergonomic eyecup facilitates eye opening.
  • Soft and autonomous rinsing without pressure
  • Sealed to assure the integrity of the system
  • Waterproof and compact EN 15454-4. EC certification
Specifications
Product Type Eyewash Solution
Sterility Sterile
Volume (Metric) 50 mL