Learn More
Diphenylmethyl 7Beta-Amino-3-cephem-4-carboxylate, TRC
Specifications
Specifications
| Chemical Name or Material | Diphenylmethyl 7beta-Amino-3-cephem-4-carboxylate |
| CAS | 36923-21-4 |
| Molecular Formula | C20H18N2O3S |
| Quantity | 500 mg |
| Synonym | (6R,7R)-7-Amino-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid Diphenylmethyl Ester, (6R-trans)-7-Amino-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid Diphenylmethyl Ester |
| InChI Formula | InChI=1S/C20H18N2O3S/c21-16-18(23)22-15(11-12-26-19(16)22)20(24)25-17(13-7-3-1-4-8-13)14-9-5-2-6-10-14/h1-11,16-17,19H,12,21H2 |
| SMILES | NC1C2SCC=C(N2C1=O)C(=O)OC(c3ccccc3)c4ccccc4 |
| IUPAC Name | benzhydryl 7-amino-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate |
| Molecular Weight (g/mol) | 366.43 |
| Formula Weight | 366.1038 |