Learn More
Diphenhydramine N-Beta-D-Glucuronide, TRC
Product Code. 30004376
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30004376 2.5 mg 2.5mg
30012567 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30004376 Supplier Toronto Research Chemicals Supplier No. TRCD4869102.5MG
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 137908-78-2
Molecular Formula C23 H29 N O7
Molecular Weight (g/mol) 431.48
Synonym Ethanaminium, 2-(diphenylmethoxy)-N-β-D-glucopyranuronosyl-N,N-dimethyl-, inner salt, Diphenhydramine N-glucuronide, Diphenhydramine N-β-D-Glucuronide
IUPAC Name (2S,3S,4S,5R,6R)-6-[2-benzhydryloxyethyl(dimethyl)azaniumyl]-3,4,5-trihydroxyoxane-2-carboxylate
SMILES C[N+](C)(CCOC(c1ccccc1)c2ccccc2)[C@@H]3O[C@@H]([C@@H](O)[C@H](O)[C@H]3O)C(=O)[O-]
Specifications
Quantity 2.5 mg
InChI Formula InChI=1S/C23H29NO7/c1-24(2,22-19(27)17(25)18(26)21(31-22)23(28)29)13-14-30-20(15-9-5-3-6-10-15)16-11-7-4-8-12-16/h3-12,17-22,25-27H,13-14H2,1-2H3/t17-,18-,19+,21-,22+/m0/s1
Formula Weight 431.1944
Chemical Name or Material Diphenhydramine N-beta-D-Glucuronide